siRNA Details
Gene Symbol: Il9r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017021.1 | NP_058717.1 | 3 | 164 | ||
XM_006245901.2 | XP_006245963.1 | 2 | 506 | |||
XM_006245903.2 | XP_006245965.1 | 3 | 526 | |||
GenBank mRNA | L36459.1 | - | 3 | 164 |
