siRNA Details
Gene Symbol: Htr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013148.1 | NP_037280.1 | 2 | 1180 | ||
GenBank mRNA | L10072.1 | - | 2 | 1180 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
