siRNA Details
Gene Symbol: Mep1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013183.2 | NP_037315.1 | 5 | 218 | ||
XM_006254458.2 | XP_006254520.1 | 4 | 359 | |||
XM_006254459.2 | XP_006254521.1 | 5 | 187 | |||
GenBank mRNA | BC081833.1 | - | 5 | 194 | 7108543 | |
M88601.1 | - | 5 | 219 |
