siRNA Details
Gene Symbol: Ppara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013196.1 | NP_037328.1 | 4 | 709 | ||
XM_006242151.2 | XP_006242213.1 | 3 | 1665 | |||
XM_006242152.2 | XP_006242214.1 | 5 | 937 | |||
XM_006242153.2 | XP_006242215.1 | 5 | 652 | |||
XM_006242154.2 | XP_006242216.1 | 5 | 706 | |||
XM_006242156.2 | XP_006242218.1 | 4 | 536 | |||
XM_008765676.1 | XP_008763898.1 | 5 | 729 | |||
GenBank mRNA | M88592.1 | - | 4 | 709 |
