siRNA Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 5 | 563 | ||
GenBank mRNA | AB017801.1 | - | 5 | 529 | ||
BC059110.1 | - | 5 | 564 | 6919611 | ||
BC087743.1 | - | 5 | 532 | 7321006 | ||
DQ403053.1 | - | 3 | 419 | |||
M17701.1 | - | 4 | 518 | |||
X02231.1 | - | 5 | 559 |
Gene Symbol: Gapdh-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC087069.1 | - | 2 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
