siRNA Details
Gene Symbol: Gsta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017013.4 | NP_058709.2 | 5 | 451 | ||
XM_006243421.2 | XP_006243483.1 | 5 | 518 | |||
XM_006243422.2 | XP_006243484.1 | 5 | 512 | |||
GenBank mRNA | BC078706.1 | - | 5 | 443 | 7128611 | |
FJ179397.1 | - | 4 | 388 | |||
K01931.1 | - | 5 | 451 | |||
M26874.1 | - | 1 | 121 |
Gene Symbol: Gsta5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010921.1 | NP_001010921.1 | 3 | 414 | ||
GenBank mRNA | BC098898.1 | - | 3 | 437 | 7322988 | |
CO566028.1 | - | 435 | ||||
K00136.1 | - | 3 | 427 | |||
M25891.1 | - | 3 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
