siRNA Details
Gene Symbol: Itgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017022.2 | NP_058718.2 | 4 | 431 | ||
XM_006255824.1 | XP_006255886.1 | 4 | 461 | |||
GenBank mRNA | BC131845.1 | - | 4 | 431 | 8366766 | |
U12309.1 | - | 4 | 305 |
