siRNA Details
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017024.2 | NP_058720.2 | 2 | 252 | ||
XM_006255441.1 | XP_006255503.1 | 2 | 721 | |||
GenBank mRNA | BC091155.1 | - | 2 | 252 | 7370790 | |
FQ209624.1 | - | 2 | 267 | |||
FQ218245.1 | - | 2 | 267 | |||
FQ218883.1 | - | 2 | 270 | |||
U62803.1 | - | 2 | 255 | |||
X54096.1 | - | 2 | 266 |
