siRNA Details
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017025.1 | NP_058721.1 | 3 | 288 | ||
XM_006229232.2 | XP_006229294.1 | 3 | 456 | |||
GenBank mRNA | BC084698.1 | - | 2 | 190 | 7122339 | |
BC168737.1 | - | 3 | 287 | 6917937 | ||
FQ214854.1 | - | 3 | 288 | |||
X01964.1 | - | 3 | 288 |
