siRNA Details
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013143.1 | NP_037275.1 | 12 | 1714 | ||
XM_008766837.1 | XP_008765059.1 | 12 | 1753 | |||
XM_008766838.1 | XP_008765060.1 | 10 | 1575 | |||
GenBank mRNA | BC081834.1 | - | 12 | 1729 | 7128404 | |
S43408.1 | - | 12 | 1714 |
