siRNA Details
Gene Symbol: Vldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013155.2 | NP_037287.2 | 10 | 1568 | ||
XM_006231197.2 | XP_006231259.1 | 11 | 1873 | |||
XM_006231198.2 | XP_006231260.1 | 11 | 1602 | |||
XM_006231199.2 | XP_006231261.1 | 10 | 1476 | |||
GenBank mRNA | L35767.1 | - | 10 | 1568 |

More Information
Other Species Targeted

Gene Ontology Categories:
