siRNA Details
Gene Symbol: Ide | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013159.1 | NP_037291.1 | 5 | 736 | ||
XM_006231313.2 | XP_006231375.1 | 5 | 1507 | |||
XM_006231314.2 | XP_006231376.1 | 5 | 720 | |||
XM_006231315.2 | XP_006231377.1 | 5 | 790 | |||
XM_008760362.1 | XP_008758584.1 | 5 | 1477 | |||
GenBank mRNA | X67269.1 | - | 5 | 736 |
