siRNA Details
Gene Symbol: Cadps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013219.1 | NP_037351.1 | 3212 | |||
GenBank mRNA | U16802.1 | - | 3212 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001042617, NM_012061, AK133310, AK161919, AK122453, and others.
Gene Ontology Categories:
