siRNA Details
Gene Symbol: Erbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017003.2 | NP_058699.2 | 19 | 2384 | ||
GenBank mRNA | AY116182.1 | - | 19 | 2384 | ||
BC061863.1 | - | 19 | 2456 | 5597835 | ||
X03362.1 | - | 19 | 2298 |
Gene Symbol: LOC102552659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247245.2 | XP_006247307.2 | 6 | 647 |
