siRNA Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 6 | 734 | ||
GenBank mRNA | AB017801.1 | - | 6 | 700 | ||
BC059110.1 | - | 6 | 735 | 6919611 | ||
BC087743.1 | - | 6 | 703 | 7321006 | ||
DQ403053.1 | - | 4 | 590 | |||
M17701.1 | - | 5 | 689 | |||
X02231.1 | - | 6 | 730 |
Gene Symbol: LOC303448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037190.1 | NP_001032267.1 | 2 | 772 | ||
GenBank mRNA | BC100150.1 | - | 2 | 772 | 7455257 |

More Information
Other Species Targeted

Gene Ontology Categories:
