siRNA Details
Gene Symbol: Kcnj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017023.1 | NP_058719.1 | 2 | 1160 | ||
GenBank mRNA | AF081365.1 | - | 3 | 984 | ||
AF081366.1 | - | 3 | 933 | |||
AF081367.1 | - | 4 | 951 | |||
AF081368.1 | - | 4 | 1106 | |||
L29403.1 | - | 2 | 1109 | |||
X72341.1 | - | 2 | 1160 |
