siRNA Details
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017024.2 | NP_058720.2 | 5 | 546 | ||
XM_006255441.1 | XP_006255503.1 | 5 | 1015 | |||
GenBank mRNA | BC091155.1 | - | 5 | 546 | 7370790 | |
FQ209624.1 | - | 5 | 561 | |||
FQ218245.1 | - | 5 | 561 | |||
FQ218883.1 | - | 5 | 564 | |||
U62803.1 | - | 5 | 549 | |||
X54096.1 | - | 5 | 560 |
