siRNA Details
Gene Symbol: STK17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004226.3 | NP_004217.1 | 2, 3 | 456 | ||
XM_011512168.2 | XP_011510470.1 | 2, 3 | 421 | |||
XM_011512169.1 | XP_011510471.1 | 2, 3 | 454 | |||
XM_011512170.1 | XP_011510472.1 | 3, 4 | 437 | |||
GenBank mRNA | AB011421.1 | - | 2, 3 | 393 | ||
CR542210.1 | - | 1, 2 | 132 | |||
CR542225.1 | - | 1, 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
