siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145842.1 | NP_001139314.1 | 3, 4 | 328 | ||
NM_017113.2 | NP_058809.2 | 3, 4 | 328 | |||
XM_008767981.1 | XP_008766203.1 | 3, 4 | 409 | |||
GenBank mRNA | BC072469.1 | - | 3, 4 | 322 | 7098196 | |
CB731349.1 | - | 3, 4 | 359 | |||
CK365572.1 | - | 3, 4 | 321 | |||
M97750.1 | - | 2, 3 | 286 | |||
X62322.1 | - | 3, 4 | 302 |
