siRNA Details
Gene Symbol: Plcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017035.1 | NP_058731.1 | 3 | 509 | ||
XM_006244063.2 | XP_006244125.1 | 3 | 408 | |||
GenBank mRNA | EF089258.1 | - | 3 | 287 | ||
M20637.1 | - | 3 | 509 |
