siRNA Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017075.2 | NP_058771.2 | 3, 4 | 321 | ||
GenBank mRNA | D00512.1 | - | 3, 4 | 253 | ||
D13921.1 | - | 3, 4 | 253 | |||
FM065829.1 | - | 3, 4 | 321 | |||
FM071064.1 | - | 1, 2 | 109 | |||
FQ217320.1 | - | 3, 4 | 259 | |||
FQ228108.1 | - | 3, 4 | 308 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144784, AK032097, AK075593, AK163402, AK081715, and others.
Gene Ontology Categories:
