siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017030.2 | NP_058726.2 | 13 | 1328 | ||
GenBank mRNA | BC078855.1 | - | 13 | 1328 | 7108495 | |
M14634.1 | - | 13 | 1329 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025835, AK050366, AF327060, AK004976, AK134437.
Gene Ontology Categories:
