siRNA Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017075.2 | NP_058771.2 | 10, 11 | 1092 | ||
GenBank mRNA | D00512.1 | - | 10, 11 | 1024 | ||
D13921.1 | - | 10, 11 | 1024 | |||
FM098848.1 | - | 5, 6 | 474 | |||
FQ217320.1 | - | 10, 11 | 1030 | |||
FQ228108.1 | - | 10, 11 | 1078 |

More Information
Other Species Targeted

Gene Ontology Categories:
