siRNA Details
Gene Symbol: Chrna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017078.2 | NP_058774.2 | 5, 6 | 1307 | ||
GenBank mRNA | AY574255.1 | - | 5, 6 | 1205 | ||
J05231.1 | - | 5, 6 | 1306 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_176844, DQ318788, AY574264, BC106839, BC106853, and others.
Gene Ontology Categories:
