siRNA Details
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017117.1 | NP_058813.1 | 13 | 1875 | ||
XM_006234728.2 | XP_006234790.1 | 13 | 1609 | |||
XM_006234729.2 | XP_006234791.1 | 13 | 1603 | |||
XM_008762080.1 | XP_008760302.1 | 13 | 2507 | |||
XM_008762081.1 | XP_008760303.1 | 13 | 2507 | |||
GenBank mRNA | AF052540.1 | - | 12 | 1383 | ||
AF061726.2 | - | 13 | 1580 | |||
AF173834.1 | - | 12 | 2149 | |||
AF184950.1 | - | 13 | 1520 | |||
J05121.1 | - | 13 | 1875 | |||
U96367.1 | - | 12 | 1432 |
