siRNA Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017196.3 | NP_058892.1 | 2 | 174 | ||
XM_006256061.2 | XP_006256123.1 | 2 | 269 | |||
XM_006256062.2 | XP_006256124.1 | 3 | 246 | |||
XM_006256063.2 | XP_006256125.1 | 3 | 209 | |||
XM_006256065.2 | XP_006256127.1 | 4 | 324 | |||
GenBank mRNA | AB000818.1 | - | 2 | 172 | ||
BC081822.1 | - | 1 | 742 | |||
D82069.1 | - | 4 | 329 | |||
FM053942.1 | - | 2 | 161 | |||
FQ231499.1 | - | 2 | 172 | |||
U17919.1 | - | 2 | 146 |
