siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083586.2 | NP_001077055.2 | 2 | 401 | ||
NM_017197.3 | NP_058893.2 | 2 | 401 | |||
XM_008771861.1 | XP_008770083.1 | 2 | 280 | |||
XM_008771862.1 | XP_008770084.1 | 1 | 838 | |||
XM_008771863.1 | XP_008770085.1 | 1 | 837 | |||
XM_008771864.1 | XP_008770086.1 | 1 | 837 | |||
GenBank mRNA | AF090695.1 | - | 2 | 235 | ||
AJ010351.1 | - | 2 | 326 | |||
AJ010386.1 | - | 2 | 449 | |||
BC129096.1 | - | 2 | 227 | 8362847 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001110231, NM_001110228, NM_001110230, NM_001160292, NM_001110229, and others.
Gene Ontology Categories:
