siRNA Details
Gene Symbol: MBTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003791.3 | NP_003782.1 | 14 | 2371 | ||
XM_017023816.1 | XP_016879305.1 | 13 | 2120 | |||
XM_017023817.1 | XP_016879306.1 | 15 | 2374 | |||
GenBank mRNA | AK091212.1 | - | 3 | 1081 | ||
AK291773.1 | - | 14 | 2254 | |||
AK299067.1 | - | 13 | 1596 | |||
BC114555.1 | - | 14 | 2257 | 40054075 | ||
BC114961.1 | - | 14 | 2257 | 40054074 | ||
D42053.1 | - | 14 | 2334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
