siRNA Details
Gene Symbol: Stmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017166.1 | NP_058862.1 | 4 | 470 | ||
XM_006239123.2 | XP_006239185.1 | 4 | 468 | |||
GenBank mRNA | BC062234.1 | - | 4 | 395 | 6917958 | |
FQ211845.1 | - | 4 | 393 | |||
FQ212924.1 | - | 4 | 392 | |||
FQ213053.1 | - | 4 | 393 | |||
FQ214201.1 | - | 4 | 387 | |||
FQ214447.1 | - | 4 | 393 | |||
FQ220047.1 | - | 4 | 393 | |||
J04979.1 | - | 4 | 470 | |||
M27876.1 | - | 3 | 413 | |||
X94914.1 | - | 2 | 302 |
