siRNA Details
Gene Symbol: Wfdc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081957.1 | NP_001075426.1 | 3 | 404 | ||
GenBank mRNA | BC038870.1 | - | 3 | 333 | ||
BC089618.1 | - | 3 | 377 | 6766535 | ||
FJ007372.1 | - | 3 | 445 | |||
W08116.1 | - | 3 | 404 |
