siRNA Details
Gene Symbol: Fam174b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162532.1 | NP_001156004.1 | 3 | 2637 | ||
GenBank mRNA | AK185650.1 | - | 1 | 86 | ||
BC034069.1 | - | 3 | 2515 | |||
BC110691.1 | - | 3 | 2352 |
