siRNA Details
Gene Symbol: Gm14920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102665.1 | NP_001096135.1 | 1 | 14 |
Gene Symbol: H2afb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281531.1 | NP_001268460.1 | 1 | 14 |
