siRNA Details
Gene Symbol: Iqcf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101628.1 | NP_001095098.1 | 2 | 448 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109362.
Gene Ontology Categories:
