siRNA Details
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029701.1 | - | 14 | 2647 | ||
AK129468.1 | - | 5175 | ||||
AK154663.1 | - | 20 | 3823 | |||
AK159547.1 | - | 4683 | ||||
AK195963.1 | - | 1 | 38 | |||
BC025835.1 | - | 2017 | ||||
BC036324.1 | - | 11 | 2867 | |||
BC092227.1 | - | 21 | 4468 | 30531788 |
