siRNA Details
Gene Symbol: Krtap10-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135991.1 | NP_001129463.1 | 1 | 868 | ||
GenBank mRNA | AK020700.1 | - | 1 | 896 | ||
BC115499.1 | - | 1 | 868 | 40100932 | ||
BC115500.1 | - | 1 | 868 | 40100933 |
