siRNA Details
Gene Symbol: Gm17359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142953.1 | NP_001136425.1 | 2 | 299 | ||
GenBank mRNA | BU583367.1 | - | 1 | 66 | ||
CN840131.1 | - | 302 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm17359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142953.1 | NP_001136425.1 | 2 | 299 | ||
GenBank mRNA | BU583367.1 | - | 1 | 66 | ||
CN840131.1 | - | 302 |