siRNA Details
Gene Symbol: Gm17677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167588.1 | NP_001161060.1 | 2, 3 | 194 | ||
GenBank mRNA | EU703628.1 | - | 2, 3 | 194 |

More Information
Other Species Targeted

