siRNA Details
Gene Symbol: Sp3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005870.1 | - | 2 | 227 | ||
AK013593.1 | - | 2 | 186 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183265.1 | - | 227 | Chr. - - [-] on Build |
