siRNA Details
Gene Symbol: 4930412M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029606.1 | - | 2 | 750 | ||
AK089395.1 | - | 1 | 2670 | |||
AK154788.1 | - | 1 | 2668 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177098.1 | - | 750 | Chr. - - [-] on Build |
