siRNA Details
Gene Symbol: AU022751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033211.3 | NP_001028383.2 | 2 | 1233 | ||
NM_001166433.1 | NP_001159905.1 | 1 | 1596 | |||
GenBank mRNA | AK087702.1 | - | 1 | 1597 | ||
AK136005.1 | - | 2 | 1234 | |||
AK166653.1 | - | 2 | 1233 | |||
BC100377.1 | - | 1 | 1582 | 30938251 |
