siRNA Details
Gene Symbol: Synj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045515.1 | NP_001038980.1 | 33 | 6808 | ||
NM_001164483.1 | NP_001157955.1 | 32 | 6877 | |||
GenBank mRNA | AB093272.1 | - | 32 | 6694 |
