siRNA Details
Gene Symbol: Synj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045515.1 | NP_001038980.1 | 28 | 3637 | ||
NM_001164483.1 | NP_001157955.1 | 28 | 3730 | |||
GenBank mRNA | AB093272.1 | - | 27 | 3523 | ||
BU703932.1 | - | 7 | 680 | |||
BY267025.1 | - | 4 | 250 |
