siRNA Details
Gene Symbol: Smim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135577.2 | NP_001129049.1 | 2 | 4037 | ||
GenBank mRNA | AK076140.1 | - | 1 | 1512 | ||
BC066054.1 | - | 2 | 4020 | |||
BC094222.1 | - | 2 | 4019 |
