siRNA Details
Gene Symbol: C330006A16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256521.1 | NP_001243450.1 | 2 | 2268 | ||
NM_001256522.1 | NP_001243451.1 | 2 | 2108 | |||
GenBank mRNA | AK077073.1 | - | 1 | 1287 | ||
AK082757.1 | - | 1 | 1290 | |||
BC020361.1 | - | 1 | 1228 |
