siRNA Details
Gene Symbol: Flna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290421.1 | NP_001277350.1 | 10, 11 | 1784 | ||
NM_010227.3 | NP_034357.2 | 10, 11 | 1786 | |||
GenBank mRNA | AK089311.1 | - | 10, 11 | 1664 | ||
AK152579.1 | - | 10, 11 | 1442 | |||
BC038478.1 | - | 10, 11 | 1659 | |||
BC151024.1 | - | 10, 11 | 1777 | 9087937 |
