siRNA Details
Gene Symbol: Flna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290421.1 | NP_001277350.1 | 33 | 5581 | ||
NM_010227.3 | NP_034357.2 | 32 | 5559 | |||
GenBank mRNA | AK131179.1 | - | 20 | 3400 | ||
BC038478.1 | - | 33 | 5456 | |||
BC039208.1 | - | 12 | 1771 | |||
BC151024.1 | - | 32 | 5550 | 9087937 |
