siRNA Details
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017274.1 | NP_058970.1 | 3 | 361 | ||
XM_006231625.2 | XP_006231687.1 | 3 | 752 | |||
XM_006231626.2 | XP_006231688.1 | 4 | 633 | |||
XM_008760534.1 | XP_008758756.1 | 3 | 303 | |||
GenBank mRNA | AF021348.1 | - | 3 | 361 | ||
U36771.2 | - | 2 | 137 |
