siRNA Details
Gene Symbol: Psma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017279.1 | NP_058975.1 | 3 | 193 | ||
GenBank mRNA | BC060576.1 | - | 3 | 212 | 6919255 | |
FQ214776.1 | - | 3 | 208 | |||
FQ223872.1 | - | 208 | ||||
J02897.1 | - | 3 | 193 |
