siRNA Details
Gene Symbol: Iglon5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164518.1 | NP_001157990.1 | 8 | 1519 | ||
GenBank mRNA | AK039193.1 | - | 8 | 1486 | ||
BC116421.1 | - | 5 | 1103 | |||
BC116422.1 | - | 5 | 1103 |
