siRNA Details
Gene Symbol: Zfp748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035231.3 | NP_001030308.1 | 4 | 3000 | ||
GenBank mRNA | AK132196.1 | - | 4 | 2987 | ||
AK141365.1 | - | 4 | 2983 | |||
AK220562.1 | - | 4 | 2979 | |||
BC055310.1 | - | 4 | 2986 | 30048242 |
